Assay Details
Target Gene Details
Entrez Gene ID: | 227753 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF |
Location: |
Chr.2:35256359-35307902 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gsn | NM_001206367.1 | NP_001193296.1 | ||
NM_001206368.1 | NP_001193297.1 | |||
NM_001206369.1 | NP_001193298.1 | |||
NM_146120.4 | NP_666232.2 | |||
AK076156.1 | BAC36223.1 | |||
AK089934.1 | BAC41004.1 | |||
AK135632.1 | ||||
AK143664.1 | BAE25485.1 | |||
AK146969.1 | BAE27574.1 | |||
AK148957.1 | BAE28702.1 | |||
AK150359.1 | BAE29494.1 | |||
AK151684.1 | BAE30608.1 | |||
AK153029.1 | BAE31661.1 | |||
AK166831.1 | BAE39054.1 | |||
AK166932.1 | BAE39126.1 | |||
AK167918.1 | BAE39925.1 | |||
AK168461.1 | BAE40358.1 | |||
AK168532.1 | BAE40410.1 | |||
AK168707.1 | BAE40549.1 | |||
AK168725.1 | BAE40566.1 | |||
AK168829.1 | ||||
AK169041.1 | BAE40830.1 | |||
AK169670.1 | BAE41291.1 | |||
AK206117.1 | ||||
AK213866.1 | ||||
AK217606.1 | ||||
AK218612.1 | ||||
AK219663.1 | ||||
BC023143.1 | ||||
BC060377.1 | AAH60377.1 | |||
J04953.1 | AAA37677.1 |
More Information
Set Membership: |
Intragenic Non-exonic |