Assay Details
Target Gene Details
Entrez Gene ID: | 107435 |
Gene Name: | histone aminotransferase 1 |
Gene Aliases: |
2410071B14Rik, AA536933, Hat-1, KAT1 |
Location: |
Chr.2:71389260-71441622 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hat1 | NM_026115.4 | 5 | 418 | NP_080391.2 |
AK010715.1 | 5 | 427 | ||
AK014330.1 | 5 | 430 | ||
AK018270.1 | 5 | 413 | ||
AK041700.1 | 5 | 422 | BAC31038.1 | |
AK137648.1 | 5 | 436 | ||
AK169315.1 | 5 | 421 | BAE41070.1 | |
AK182724.1 | 4 | 376 | ||
BC055460.1 | 4 | 385 | AAH55460.1 |
More Information
Set Membership: |
Intragenic Exonic |