Assay Details
Target Gene Details
Entrez Gene ID: | 74533 |
Gene Name: | GDNF-inducible zinc finger protein 1 |
Gene Aliases: |
8430437G08Rik, Zfp336, mGZF1 |
Location: |
Chr.2:148679385-148692949 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gzf1 | NM_028986.3 | NP_083262.1 | ||
AB100266.1 | BAC98465.1 | |||
AK018470.1 | BAB31226.1 | |||
AK052820.1 | BAC35160.1 | |||
AK053262.1 | BAC35327.1 | |||
AK141153.1 | BAE24571.1 | |||
BC096015.1 | AAH96015.1 |
More Information
Set Membership: |
Intragenic Non-exonic |