Assay Details
Target Gene Details
Entrez Gene ID: | 14571 |
Gene Name: | glycerol phosphate dehydrogenase 2, mitochondrial |
Gene Aliases: |
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38 |
Location: |
Chr.2:57237678-57370719 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpd2 | NM_001145820.1 | 12 | 1865 | NP_001139292.1 |
NM_010274.3 | 12 | 1740 | NP_034404.3 | |
AK034353.1 | 12 | 1649 | BAC28685.1 | |
AK144716.1 | 12 | 1866 | BAE26028.1 | |
AK151189.1 | 12 | 1653 | ||
AK167152.1 | 12 | 1678 | BAE39294.1 | |
BC021359.1 | 12 | 1834 | AAH21359.1 | |
D50430.1 | 12 | 1642 | BAA08926.1 | |
U60987.1 | 12 | 1676 | AAB50545.1 |
More Information
Set Membership: |
Intragenic Exonic |