Assay Details
Target Gene Details
Entrez Gene ID: | 14571 |
Gene Name: | glycerol phosphate dehydrogenase 2, mitochondrial |
Gene Aliases: |
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38 |
Location: |
Chr.2:57237678-57370719 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpd2 | NM_001145820.1 | NP_001139292.1 | ||
NM_010274.3 | NP_034404.3 | |||
AK034353.1 | BAC28685.1 | |||
AK144716.1 | BAE26028.1 | |||
AK151189.1 | ||||
AK167152.1 | BAE39294.1 | |||
BC021359.1 | AAH21359.1 | |||
BY238564.1 | ||||
D50430.1 | BAA08926.1 | |||
U60987.1 | AAB50545.1 |
More Information
Set Membership: |
Intragenic Non-exonic |