Assay Details
Target Gene Details
Entrez Gene ID: | 14425 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 |
Gene Aliases: |
- |
Location: |
Chr.2:66082766-66124793 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt3 | NM_015736.2 | 7 | 1679 | NP_056551.2 |
AK019995.1 | 7 | 1678 | ||
AK135489.1 | 7 | 1640 | BAE22551.1 | |
BC043331.1 | 7 | 1535 | AAH43331.1 | |
U70538.1 | 7 | 1523 | AAB09579.1 |
More Information
Set Membership: |
Intragenic Exonic |