Assay Details
Target Gene Details
Entrez Gene ID: | 271786 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 |
Gene Aliases: |
A230002A12, A230020F20, BB182356 |
Location: |
Chr.2:54436387-55118309 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt13 | NM_173030.2 | 3 | 526 | NP_766618.2 |
AB082928.1 | BAC54546.1 | |||
AK030726.1 | 2 | 456 | ||
AK038387.1 | 3 | 527 | BAC29981.1 | |
AK045417.1 | 3 | 527 | BAC32353.1 | |
BC131651.1 | 3 | 205 | ||
BC131652.1 | 2 | 236 |
More Information
Set Membership: |
Intragenic Exonic |