Assay Details
Target Gene Details
Entrez Gene ID: | 14415 |
Gene Name: | glutamate decarboxylase 1 |
Gene Aliases: |
EP10, GAD25, GAD44, GAD67, Gad-1 |
Location: |
Chr.2:70561763-70602014 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gad1 | NM_008077.4 | NP_032103.2 | ||
AF326547.1 | AAL37393.1 | |||
AF326548.1 | AAL37394.1 | |||
AF483492.1 | AAL90766.1 | |||
AF483493.1 | AAL90767.1 | |||
AK047521.1 | ||||
AK054554.1 | BAC35823.1 | |||
AK136801.1 | BAE23133.1 | |||
AK148497.1 | BAE28589.1 | |||
AK160410.1 | ||||
AK160868.1 | BAE36059.1 | |||
BC027059.1 | AAH27059.1 | |||
KC134210.1 | ||||
KM102984.1 | ||||
KM102985.1 | ||||
Y12257.1 | CAA72934.1 | |||
Z49976.1 | CAA90277.1 |
More Information
Set Membership: |
Intragenic Non-exonic |