Assay Details
Target Gene Details
Entrez Gene ID: | 14378 |
Gene Name: | glucose-6-phosphatase, catalytic, 2 |
Gene Aliases: |
G6pc-rs, IGRP |
Location: |
Chr.2:69211073-69227993 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G6pc2 | NM_001289856.1 | 3 | 250 | NP_001276785.1 |
NM_001289857.1 | 3 | 470 | NP_001276786.1 | |
NM_021331.4 | 3 | 470 | NP_067306.1 | |
AK148450.1 | 3 | 471 | ||
AK148465.1 | 3 | 422 | BAE28569.1 | |
BC111905.1 | 3 | 445 | AAI11906.1 | |
BQ557220.1 | 3 | 250 | ||
Z47787.1 | 3 | 448 | CAA87708.1 |
More Information
Set Membership: |
Intragenic Exonic |