Assay Details
Target Gene Details
Entrez Gene ID: | 654821 |
Gene Name: | glucosaminyl (N-acetyl) transferase family member 7 |
Gene Aliases: |
A330041C17Rik, gnt |
Location: |
Chr.2:172447120-172458596 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gcnt7 | NM_001039560.3 | NP_001034649.1 | ||
AK039414.1 | BAE20545.1 | |||
AK132647.1 | BAE21278.1 | |||
BC139073.1 | ||||
BC139076.1 |
Target Gene Details
Entrez Gene ID: | 66404 |
Gene Name: | replication termination factor 2 domain containing 1 |
Gene Aliases: |
1700067C04Rik, 2410001C21Rik, 5730427M17Rik, AA589417 |
Location: |
Chr.2:172440578-172469899 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rtfdc1 | NM_025542.2 | NP_079818.1 | ||
AK009901.1 | BAB26573.1 | |||
AK010321.1 | ||||
AK028792.1 | BAC26123.1 | |||
AK053258.1 | BAC35324.1 | |||
AK077823.1 | BAC37024.1 | |||
AK153121.1 | BAE31736.1 | |||
AK167287.1 | BAE39393.1 | |||
AK169588.1 | BAE41244.1 | |||
AK170141.1 | BAE41592.1 | |||
BC004803.1 | AAH04803.1 | |||
BC049072.1 | AAH49072.2 | |||
BC096692.1 | AAH96692.1 | |||
BC116442.1 | ||||
BC116443.1 | ||||
BC127166.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |