Assay Details
Target Gene Details
Entrez Gene ID: | 71409 |
Gene Name: | formin-like 2 |
Gene Aliases: |
5430425K04Rik, Man |
Location: |
Chr.2:52857716-53134202 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fmnl2 | NM_172409.2 | 22 | 3039 | NP_765997.2 |
AF513716.1 | 11 | 1436 | AAP87551.1 | |
AK080318.1 | 2 | 214 | BAC37877.1 | |
AK173293.1 | 5 | 386 | BAD32571.1 | |
BC048004.1 | 7 | 927 | AAH48004.1 | |
BC064731.1 | 2 | 196 | AAH64731.1 |