Assay Details
Target Gene Details
Entrez Gene ID: | 227715 |
Gene Name: | exosome component 2 |
Gene Aliases: |
Rrp4 |
Location: |
Chr.2:31670737-31681349 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Exosc2 | NM_144886.2 | NP_659135.1 | ||
AK144234.1 | BAE25788.1 | |||
AK164618.1 | ||||
AK166881.1 | BAE39092.1 | |||
BC021485.1 | AAH21485.1 | |||
BC021807.1 | AAH21807.1 |
More Information
Set Membership: |
Intragenic Non-exonic |