Assay Details
Target Gene Details
Entrez Gene ID: | 14089 |
Gene Name: | fibroblast activation protein |
Gene Aliases: |
SIMP |
Location: |
Chr.2:62500936-62575508 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fap | NM_007986.3 | 20 | 1928 | NP_032012.1 |
BC019190.1 | 20 | 1832 | AAH19190.1 | |
Y10007.1 | 20 | 1928 | CAA71116.1 |
More Information
Set Membership: |
Intragenic Exonic |