Assay Details
Target Gene Details
Entrez Gene ID: | 14089 |
Gene Name: | fibroblast activation protein |
Gene Aliases: |
SIMP |
Location: |
Chr.2:62500936-62575508 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fap | NM_007986.3 | NP_032012.1 | ||
AK051959.1 | ||||
AK136578.1 | ||||
BC019190.1 | AAH19190.1 | |||
Y10007.1 | CAA71116.1 |
More Information
Set Membership: |
Intragenic Non-exonic |