Assay Details
Target Gene Details
Entrez Gene ID: | 353156 |
Gene Name: | EGF-like domain 7 |
Gene Aliases: |
VE-statin, Zneu1 |
Location: |
Chr.2:26581028-26592682 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egfl7 | NM_178444.4 | NP_848538.3 | ||
NM_198724.2 | 1 | 97 | NP_942017.2 | |
NM_198725.2 | 1 | 97 | NP_942018.2 | |
AF184973.1 | AAF01322.1 | |||
AK002601.1 | 1 | 93 | BAB22222.1 | |
AK156158.1 | ||||
AY239289.1 | AAP69825.1 | |||
AY239290.1 | 1 | 81 | AAP69826.1 | |
AY309459.1 | AAP74732.1 | |||
BC024610.1 | 1 | 81 | AAH24610.2 | |
BY274501.1 | 1 | 97 |
More Information
Set Membership: |
Intragenic Exonic |