Assay Details
Target Gene Details
Entrez Gene ID: | 320132 |
Gene Name: | RIKEN cDNA A230108P19 gene |
Gene Aliases: |
- |
Location: |
Chr.2:6193251-6321611 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A230108P19Rik | NR_040333.1 | |||
AK039218.1 | ||||
AK044786.1 |
Target Gene Details
Entrez Gene ID: | 67856 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 3 |
Gene Aliases: |
2310005D12Rik, AI662097 |
Location: |
Chr.2:6188465-6212994 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Echdc3 | NM_024208.4 | NP_077170.2 | ||
AK004218.1 | ||||
AK009166.1 | BAB26117.1 | |||
AK014736.1 | ||||
BC002214.1 | AAH02214.1 | |||
BC054365.1 | AAH54365.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |