Assay Details
Target Gene Details
Entrez Gene ID: | 110074 |
Gene Name: | deoxyuridine triphosphatase |
Gene Aliases: |
5031412I06Rik, 5133400F09Rik, D2Bwg0749e, Dutp, dUTPase |
Location: |
Chr.2:125247248-125259049 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dut | NM_001159646.1 | 1 | 82 | NP_001153118.1 |
AK017216.1 | 1 | 73 | ||
AK135438.1 | 1 | 91 | BAE22532.1 | |
BC019979.1 | 1 | 64 | AAH19979.1 |
More Information
Set Membership: |
Intragenic Exonic |