Assay Details
Target Gene Details
Entrez Gene ID: | 13482 |
Gene Name: | dipeptidylpeptidase 4 |
Gene Aliases: |
Cd26, Dpp-4, THAM |
Location: |
Chr.2:62330073-62412317 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpp4 | NM_001159543.1 | 9 | 1127 | NP_001153015.1 |
NM_010074.3 | 9 | 1127 | NP_034204.1 | |
AK040409.1 | 8 | 833 | ||
AK085370.1 | 9 | 710 | BAC39434.1 | |
AK153939.1 | 9 | 741 | BAE32266.1 | |
AK163085.1 | 9 | 1132 | BAE37187.1 | |
AK164775.1 | 9 | 710 | ||
BC022183.1 | 9 | 989 | AAH22183.1 | |
X58384.1 | 9 | 804 | CAA41274.1 |
More Information
Set Membership: |
Intragenic Exonic |