Assay Details
Target Gene Details
Entrez Gene ID: | 13482 |
Gene Name: | dipeptidylpeptidase 4 |
Gene Aliases: |
Cd26, Dpp-4, THAM |
Location: |
Chr.2:62330073-62412317 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpp4 | NM_001159543.1 | NP_001153015.1 | ||
NM_010074.3 | NP_034204.1 | |||
AK085370.1 | BAC39434.1 | |||
AK153939.1 | BAE32266.1 | |||
AK163085.1 | BAE37187.1 | |||
AK164775.1 | ||||
BC022183.1 | AAH22183.1 | |||
X58384.1 | CAA41274.1 |
More Information
Set Membership: |
Intragenic Non-exonic |