Assay Details
Target Gene Details
Entrez Gene ID: | 13436 |
Gene Name: | DNA methyltransferase 3B |
Gene Aliases: |
MmuIIIB |
Location: |
Chr.2:153649165-153687730 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnmt3b | NM_001003960.4 | NP_001003960.2 | ||
NM_001003961.4 | NP_001003961.2 | |||
NM_001003963.4 | NP_001003963.2 | |||
NM_001122997.2 | NP_001116469.1 | |||
NM_001271744.1 | NP_001258673.1 | |||
NM_001271745.1 | NP_001258674.1 | |||
NM_001271746.1 | NP_001258675.1 | |||
NM_001271747.1 | NP_001258676.1 | |||
NM_010068.5 | NP_034198.3 | |||
AF068626.2 | AAC40178.2 | |||
AF068627.2 | AAC40179.2 | |||
AF068628.2 | AAC40180.2 | |||
AF151969.1 | AAF74515.1 | |||
AF151970.1 | AAF74516.1 | |||
AF151971.1 | AAF74517.1 | |||
AF151972.1 | AAF74518.1 | |||
AF151973.1 | AAF74519.1 | |||
AF151974.1 | AAF74520.1 | |||
AF151975.1 | AAF74521.1 | |||
AF151976.1 | AAF74522.1 | |||
AY078427.1 | AAL85481.1 | |||
BC105677.1 | ||||
BC105922.1 | AAI05923.1 |
More Information
Set Membership: |
Intragenic Non-exonic |