Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dab2ip | NM_001114124.2 | 3 | 471 | NP_001107596.1 |
NM_001114125.1 | 3 | 323 | NP_001107597.1 | |
NM_001290637.1 | 3 | 378 | NP_001277566.1 | |
NM_001290641.1 | 3 | 604 | NP_001277570.1 | |
AK147464.1 | 3 | 403 | BAE27930.1 | |
AK147593.1 | 3 | 605 | BAE28013.1 | |
AK155296.1 | 3 | 379 | BAE33173.1 | |
AK164475.1 | 3 | 386 | BAE37801.1 | |
AY305656.1 | 1 | 64 | AAQ77379.1 | |
DQ473307.1 | 3 | 208 |
More Information
Set Membership: |
Intragenic Exonic |