Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dab2ip | NM_001114125.1 | NP_001107597.1 |
Target Gene Details
Entrez Gene ID: | 100042194 |
Gene Name: | predicted gene 13446 |
Gene Aliases: |
OTTMUSG00000012261 |
Location: |
Chr.2:35549534-35558702 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm13446 | NR_045894.1 | |||
AK138201.1 |
More Information
Set Membership: |
Intragenic Non-exonic |