Assay Details
Target Gene Details
Entrez Gene ID: | 12953 |
Gene Name: | cryptochrome 2 (photolyase-like) |
Gene Aliases: |
AV006279, D130054K12Rik |
Location: |
Chr.2:92403646-92434069 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cry2 | NM_009963.4 | NP_034093.1 | ||
AB003433.1 | BAA19864.1 | |||
AF156987.1 | AAD46561.1 | |||
AK037778.1 | 2 | 321 | ||
AK041696.1 | BAC31037.1 | |||
AK133781.1 | BAE21836.1 | |||
AK172994.1 | BAD32272.1 | |||
BC054794.1 | AAH54794.1 | |||
BC066799.1 | AAH66799.1 | |||
CJ164467.1 |
Target Gene Details
Entrez Gene ID: | 320507 |
Gene Name: | RIKEN cDNA D930015M05 gene |
Gene Aliases: |
- |
Location: |
Chr.2:92408892-92432308 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
D930015M05Rik | NR_040621.1 | |||
AK053012.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |