Assay Details
Target Gene Details
Entrez Gene ID: | 228094 |
Gene Name: | ceramide kinase-like |
Gene Aliases: |
Gm1958, Rp26 |
Location: |
Chr.2:79331105-79430520 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cerkl | NM_001048176.1 | 7 | 1034 | NP_001041641.1 |
CJ235603.1 | ||||
DT912627.1 | 3 | 230 | ||
HQ426680.1 | 7 | 1034 | ||
HQ426681.1 | 5 | 827 | ||
HQ426682.1 | 4 | 695 | ||
HQ426683.1 | 3 | 452 | ||
HQ426684.1 | 6 | 902 | ||
HQ426685.1 | 7 | 1034 | ||
HQ426686.1 | 6 | 891 | ||
HQ426687.1 | 7 | 1165 | ||
HQ426688.1 | 7 | 1181 | ||
HQ426689.1 | 6 | 1049 | ||
HQ426690.1 | 7 | 1251 | ||
HQ426702.1 | 6 | 616 | ||
HQ426703.1 | 4 | 409 | ||
HQ426704.1 | 6 | 616 | ||
HQ426705.1 | 5 | 473 | ||
HQ426706.1 | 5 | 560 | ||
HQ426707.1 | 5 | 707 | ||
HQ426708.1 | ||||
HQ426709.1 | 3 | 308 | ||
HQ426710.1 | 3 | 308 | ||
HQ426711.1 | 3 | 455 |
More Information
Set Membership: |
Intragenic Exonic |