Assay Details
Target Gene Details
Entrez Gene ID: | 228094 |
Gene Name: | ceramide kinase-like |
Gene Aliases: |
Gm1958, Rp26 |
Location: |
Chr.2:79331105-79430520 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cerkl | NM_001048176.1 | NP_001041641.1 | ||
DT912627.1 | ||||
HQ426680.1 | ||||
HQ426681.1 | ||||
HQ426682.1 | ||||
HQ426683.1 | ||||
HQ426684.1 | ||||
HQ426685.1 | ||||
HQ426686.1 | ||||
HQ426687.1 | ||||
HQ426688.1 | ||||
HQ426689.1 | ||||
HQ426690.1 | ||||
HQ426702.1 | ||||
HQ426703.1 | ||||
HQ426704.1 | ||||
HQ426705.1 | ||||
HQ426706.1 | ||||
HQ426707.1 | ||||
HQ426708.1 | ||||
HQ426709.1 | ||||
HQ426710.1 | ||||
HQ426711.1 |
More Information
Set Membership: |
Intragenic Non-exonic |