Assay Details
Target Gene Details
Entrez Gene ID: | 17151 |
Gene Name: | cyclin D-type binding-protein 1 |
Gene Aliases: |
AU022347, DIP1, GCIP, Maid, SECC-8, SSEC-8 |
Location: |
Chr.2:121008383-121016912 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccndbp1 | NM_010761.2 | 3 | 302 | NP_034891.2 |
AK144883.1 | 3 | 279 | BAE26115.1 | |
AK154285.1 | 3 | 256 | BAE32489.1 | |
AK160205.1 | 3 | 268 | BAE35692.1 | |
AK163150.1 | 3 | 215 | BAE37214.1 | |
AK167345.1 | 3 | 267 | BAE39444.1 | |
AK186314.1 | 2 | 79 | ||
BC001984.1 | 3 | 238 | ||
BC005802.1 | 2 | 311 | AAH05802.1 | |
CO810054.1 | 3 | 309 | ||
U50734.1 | 3 | 220 | AAB58118.1 |