Assay Details
Target Gene Details
Entrez Gene ID: | 17151 |
Gene Name: | cyclin D-type binding-protein 1 |
Gene Aliases: |
AU022347, DIP1, GCIP, Maid, SECC-8, SSEC-8 |
Location: |
Chr.2:121008383-121016912 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccndbp1 | NM_010761.2 | NP_034891.2 | ||
AK144883.1 | BAE26115.1 | |||
AK154285.1 | BAE32489.1 | |||
AK160205.1 | BAE35692.1 | |||
AK163150.1 | BAE37214.1 | |||
AK167345.1 | BAE39444.1 | |||
AK186314.1 | ||||
BC001984.1 | ||||
BC005802.1 | AAH05802.1 | |||
CO810054.1 | ||||
U50734.1 | AAB58118.1 |