Assay Details
Target Gene Details
Entrez Gene ID: | 381409 |
Gene Name: | cadherin-like 26 |
Gene Aliases: |
Gm1010 |
Location: |
Chr.2:178430515-178487366 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh26 | NM_001291189.1 | 2 | 378 | NP_001278118.1 |
NM_198656.2 | 2 | 378 | NP_941058.1 | |
AK132328.1 | 2 | 363 | BAE21107.1 | |
BC053928.1 | 2 | 286 | AAH53928.1 |
More Information
Set Membership: |
Intragenic Exonic |