Assay Details
Target Gene Details
Entrez Gene ID: | 407823 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2B |
Gene Aliases: |
5830435C13Rik, BC053917, D2Ertd794e |
Location: |
Chr.2:59899363-60209846 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz2b | NM_001001182.3 | 22 | 3616 | NP_001001182.2 |
AK029231.1 | 16 | 2664 | ||
AK047236.1 | 1 | 249 | ||
AK173180.1 | 22 | 3648 | BAD32458.1 | |
BC150814.1 | 24 | 3625 | ||
CN534605.1 | 2 | 183 |
More Information
Set Membership: |
Intragenic Exonic |