Assay Details
Target Gene Details
Entrez Gene ID: | 65964 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta |
Location: |
Chr.2:72285637-72442610 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zak | NM_001164791.1 | NP_001158263.1 | ||
NM_023057.5 | NP_075544.1 | |||
AB049731.1 | BAB16442.1 | |||
AB049732.1 | BAB16443.1 | |||
AK087363.1 | 11 | 1135 | ||
AK132186.1 | BAE21021.1 | |||
AK162819.1 | BAE37068.1 | |||
BC023718.1 | AAH23718.1 |
More Information
Set Membership: |
Intragenic Non-exonic |