Assay Details
Target Gene Details
Entrez Gene ID: | 228033 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C3 (subunit 9) |
Gene Aliases: |
6030447M23 |
Location: |
Chr.2:73908447-73911326 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp5g3 | NM_001301721.1 | 4 | 367 | NP_001288650.1 |
NM_001301722.1 | 4 | 357 | NP_001288651.1 | |
NM_175015.3 | 4 | 357 | NP_778180.1 | |
AI851824.1 | ||||
AK077930.1 | 4 | 326 | BAC37071.1 | |
AK087955.1 | 4 | 334 | BAC40056.1 | |
AW554443.2 | 2 | 313 | ||
BC116218.2 | 3 | 287 | ||
CF104340.1 | 4 | 353 | ||
CX208968.1 | 4 | 324 |
More Information
Set Membership: |
Intragenic Exonic |