Assay Details
Target Gene Details
Entrez Gene ID: | 11909 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP |
Location: |
Chr.2:73816509-73892639 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atf2 | NM_001025093.2 | 7 | 628 | NP_001020264.1 |
NM_001284369.1 | 5 | 468 | NP_001271298.1 | |
NM_001284370.1 | 7 | 628 | NP_001271299.1 | |
NM_001284371.1 | 6 | 565 | NP_001271300.1 | |
NM_001284372.1 | 7 | 628 | NP_001271301.1 | |
NM_001284373.1 | 6 | 565 | NP_001271302.1 | |
NM_001284374.1 | 5 | 468 | NP_001271303.1 | |
NM_001284376.1 | 6 | 531 | NP_001271305.1 | |
NM_009715.3 | 6 | 531 | NP_033845.1 | |
AF483482.1 | 3 | 205 | AAL90756.1 | |
AF483483.1 | 3 | 205 | AAL90757.1 | |
AK035438.1 | 7 | 584 | BAC29065.1 | |
AK041145.1 | 1 | 2206 | ||
AK047405.1 | 1 | 2009 | ||
AK051591.1 | 6 | 521 | BAC34684.1 | |
AK087424.1 | 6 | 553 | BAC39869.1 | |
AK139515.1 | 5 | 383 | ||
AK139844.1 | 5 | 381 | BAE24156.1 | |
AK140855.1 | 6 | 521 | BAE24498.1 | |
AK144977.1 | 7 | 602 | ||
AK145887.1 | 6 | 565 | BAE26726.1 | |
AK146839.1 | 6 | 477 | ||
AY902311.1 | 2 | 91 | AAX83926.1 | |
BC042210.1 | 4 | 304 | AAH42210.1 | |
BC079883.1 | 6 | 474 | AAH79883.1 | |
BC082596.1 | 6 | 511 | AAH82596.1 | |
M31629.1 | AAA39780.1 | |||
M77167.1 | AAA40394.1 |
More Information
Set Membership: |
Intragenic Exonic |