Assay Details
Target Gene Details
Entrez Gene ID: | 65964 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta |
Location: |
Chr.2:72285637-72442610 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zak | NM_023057.5 | 14 | 1389 | NP_075544.1 |
AB049731.1 | 13 | 1172 | BAB16442.1 | |
AK132186.1 | 14 | 1390 | BAE21021.1 | |
BC023718.1 | 14 | 1270 | AAH23718.1 |
More Information
Set Membership: |
Intragenic Exonic |