Assay Details
Target Gene Details
Entrez Gene ID: | 269378 |
Gene Name: | S-adenosylhomocysteine hydrolase |
Gene Aliases: |
AA987153, AL024110, CuBP, SAHH |
Location: |
Chr.2:155059310-155074497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahcy | NM_016661.3 | NP_057870.3 | ||
AK151097.1 | BAE30109.1 | |||
AK153421.1 | BAE31981.1 | |||
AK169331.1 | BAE41084.1 | |||
BC086781.1 | AAH86781.1 | |||
BC108367.1 | AAI08368.1 | |||
BQ929555.1 | ||||
L32836.1 | AAA70378.1 |
More Information
Set Membership: |
Intragenic Non-exonic |