Assay Details
Target Gene Details
Entrez Gene ID: | 11432 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
Acp-2, LAP |
Location: |
Chr.2:91202812-91214098 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acp2 | NM_007387.2 | 9 | 932 | NP_031413.1 |
AK089401.1 | 9 | 923 | ||
AK128963.1 | 9 | 925 | ||
AK133756.1 | 9 | 928 | BAE21825.1 | |
AK154269.1 | 9 | 928 | BAE32478.1 | |
AK170658.1 | 9 | 932 | ||
BC023343.1 | 9 | 914 | AAH23343.1 | |
X57199.1 | 9 | 903 | CAA40485.1 |
More Information
Set Membership: |
Intragenic Exonic |