Assay Details
Target Gene Details
Entrez Gene ID: | 76192 |
Gene Name: | abhydrolase domain containing 12 |
Gene Aliases: |
1500011G07Rik, 6330583M11Rik, AI431047, AW547313 |
Location: |
Chr.2:150832493-150904745 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd12 | NM_024465.3 | 12 | 1278 | NP_077785.2 |
AK160126.1 | 12 | 1282 | ||
AK178612.1 | 5 | 348 | ||
AK185693.1 | 5 | 326 | ||
AK198485.1 | 5 | 348 | ||
AK202769.1 | ||||
AK206881.1 | 4 | 296 | ||
AK216141.1 | 4 | 265 | ||
AK217770.1 | ||||
AK219355.1 | 5 | 350 | ||
BC002138.1 | 12 | 1247 | AAH02138.1 | |
BC002263.1 | 8 | 581 | AAH02263.1 |
More Information
Set Membership: |
Intragenic Exonic |