Assay Details
Target Gene Details
Entrez Gene ID: | 76192 |
Gene Name: | abhydrolase domain containing 12 |
Gene Aliases: |
1500011G07Rik, 6330583M11Rik, AI431047, AW547313 |
Location: |
Chr.2:150832493-150904745 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd12 | NM_024465.3 | NP_077785.2 | ||
AK160126.1 | ||||
AK178612.1 | ||||
AK185693.1 | ||||
AK198485.1 | ||||
AK202769.1 | ||||
AK206881.1 | ||||
AK216141.1 | ||||
AK217770.1 | ||||
AK219355.1 | ||||
BC002138.1 | AAH02138.1 | |||
BC002263.1 | AAH02263.1 |
More Information
Set Membership: |
Intragenic Non-exonic |