Assay Details
Target Gene Details
Entrez Gene ID: | 227736 |
Gene Name: | RIKEN cDNA 1700019L03 gene |
Gene Aliases: |
- |
Location: |
Chr.2:32777413-32784405 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700019L03Rik | NM_025619.1 | 5 | 947 | NP_079895.1 |
AK006127.1 | 5 | 971 | BAB24421.1 | |
BC118617.1 | 5 | 947 | ||
BC121825.1 | 5 | 947 |
More Information
Set Membership: |
Intragenic Exonic |