Assay Details
Target Gene Details
Entrez Gene ID: | 67388 |
Gene Name: | RIKEN cDNA 1110008F13 gene |
Gene Aliases: |
AI316789 |
Location: |
Chr.2:156863122-156873563 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1110008F13Rik | NM_026124.3 | NP_080400.1 | ||
AK002452.1 | BAB22111.1 | |||
AK003568.1 | BAB22864.1 | |||
AK078178.1 | ||||
AK184175.1 | ||||
AK189929.1 | ||||
BC019445.1 | AAH19445.1 |