Assay Details
Target Gene Details
Entrez Gene ID: | 68481 |
Gene Name: | myelin protein zero-like 1 |
Gene Aliases: |
1110007A10Rik, PZR |
Location: |
Chr.1:165592181-165634558 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpzl1 | NM_001001880.2 | NP_001001880.1 | ||
NM_001083897.1 | NP_001077366.1 | |||
AK076882.1 | ||||
AK090278.1 | ||||
AK169385.1 | BAE41132.1 | |||
AK189245.1 | ||||
AK192253.1 | ||||
AK192408.1 | ||||
AK193736.1 | ||||
AK197743.1 | ||||
AK210564.1 | ||||
AY764247.1 | AAX11678.1 | |||
AY764248.1 | AAX11679.1 | |||
BC072563.1 | AAH72563.1 |
More Information
Set Membership: |
Intragenic Non-exonic |