Assay Details
Target Gene Details
Entrez Gene ID: | 68393 |
Gene Name: | monoacylglycerol O-acyltransferase 1 |
Gene Aliases: |
0610030A14Rik, 1110064N14Rik, Dgat2l, Dgat2l1, MGAT1, WI1-2612I11.1, mDC2 |
Location: |
Chr.1:78511060-78538173 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mogat1 | NM_026713.3 | 7 | 1043 | NP_080989.2 |
AF384162.1 | 6 | 920 | AAK84177.1 | |
AK002693.1 | 6 | 992 | BAB22288.1 | |
AK004368.1 | 2 | 189 | BAE43174.1 | |
BC106135.1 | 6 | 991 | AAI06136.1 | |
BE626067.1 | 1 | 176 |
More Information
Set Membership: |
Intragenic Exonic |