Assay Details
Target Gene Details
Entrez Gene ID: | 240690 |
Gene Name: | suppression of tumorigenicity 18 |
Gene Aliases: |
AV348974, Myt3, NZF-3, Nzf3, mKIAA0535 |
Location: |
Chr.1:6487231-6860940 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
St18 | NM_001244692.1 | 20 | 3011 | NP_001231621.1 |
NM_001244693.1 | 19 | 2933 | NP_001231622.1 | |
NM_173868.2 | 18 | 2824 | NP_776293.1 | |
NR_045188.1 | 20 | 2975 | ||
NR_045189.1 | 19 | 2897 | ||
AB097467.1 | 18 | 2849 | ||
AJ536155.1 | 20 | 2975 | CAD60089.1 | |
AK122303.1 | 18 | 2773 | BAC65585.1 | |
AK137440.1 | 18 | 2774 | BAE23351.1 | |
AK147320.1 | 20 | 3012 | BAE27844.1 | |
AK147664.1 | 20 | 3012 | BAE28057.1 | |
BC032273.1 | 13 | 1738 | AAH32273.1 | |
BC118528.1 | 19 | 2847 |
More Information
Set Membership: |
Intragenic Exonic |