Assay Details
Target Gene Details
Entrez Gene ID: | 18582 |
Gene Name: | phosphodiesterase 6D, cGMP-specific, rod, delta |
Gene Aliases: |
AI841218, PrBP/delta |
Location: |
Chr.1:86543013-86582793 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde6d | NM_008801.2 | 1 | 102 | NP_032827.1 |
AF039216.1 | 1 | 61 | AAC25686.1 | |
AK141209.1 | 1 | 108 | BAE24588.1 | |
AK165701.1 | 1 | 234 | ||
AK170067.1 | 1 | 114 | BAE41543.1 | |
BC005636.1 | 1 | 102 | AAH05636.1 |
More Information
Set Membership: |
Intragenic Exonic |