Assay Details
Target Gene Details
Entrez Gene ID: | 13849 |
Gene Name: | epoxide hydrolase 1, microsomal |
Gene Aliases: |
AI195553, Eph-1, Eph1, Epxh1, mEH |
Location: |
Chr.1:180989556-181017495 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ephx1 | NM_010145.2 | NP_034275.1 | ||
AK018249.1 | BAB31132.1 | |||
AK177895.1 | ||||
AK180321.1 | ||||
AK183956.1 | ||||
AK197199.1 | ||||
AK204221.1 | ||||
AK206265.1 | ||||
AK207217.1 | ||||
AK207438.1 | ||||
AK207528.1 | ||||
AK207577.1 | ||||
AK207857.1 | ||||
AK208317.1 | ||||
AK208856.1 | ||||
AK208963.1 | ||||
AK209452.1 | ||||
AK209650.1 | ||||
AK210728.1 | ||||
AK211418.1 | ||||
AK211501.1 | ||||
AK211877.1 | ||||
AK211958.1 | ||||
AK213096.1 | ||||
AK214384.1 | ||||
AK216019.1 | ||||
AK216442.1 | ||||
AK217465.1 | ||||
AK218715.1 | ||||
AK219272.1 | ||||
AK219593.1 | ||||
BC029636.1 | AAH29636.1 | |||
BC045194.1 | AAH45194.1 | |||
BC057857.1 | AAH57857.1 | |||
BC061493.1 | AAH61493.1 | |||
U89491.1 | AAB49762.1 |
More Information
Set Membership: |
Intragenic Non-exonic |