Assay Details
Target Gene Details
Entrez Gene ID: | 67198 |
Gene Name: | spermatogenesis associated, serine-rich 2-like |
Gene Aliases: |
2810022L02Rik, A230104H11Rik, AA987173, AI480531, AI842453, AW413586 |
Location: |
Chr.1:57774162-57948398 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spats2l | NM_001164566.1 | 6 | 727 | NP_001158038.1 |
NM_144882.4 | 8 | 913 | NP_659131.2 | |
AK031266.1 | 8 | 868 | BAC27328.1 | |
AK032169.1 | 8 | 848 | BAC27738.1 | |
BC014764.1 | 6 | 727 | AAH14764.1 | |
BC021879.1 | 8 | 849 | AAH21879.1 | |
BY739337.1 | ||||
CK624383.1 | ||||
CN698528.1 | 2 | 264 |