Assay Details
Target Gene Details
Entrez Gene ID: | 319651 |
Gene Name: | ubiquitin specific peptidase 37 |
Gene Aliases: |
4932415L06Rik, C330008N13Rik |
Location: |
Chr.1:74435510-74544286 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp37 | NM_176972.4 | 5 | 656 | NP_795946.2 |
AK030013.1 | 5 | 657 | BAC26734.1 | |
AK035640.1 | 4 | 640 | BAC29135.1 | |
AK173210.1 | 1 | 132 | BAD32488.1 | |
BC139091.1 | 5 | 598 |
More Information
Set Membership: |
Intragenic Exonic |