Assay Details
Target Gene Details
Entrez Gene ID: | 22637 |
Gene Name: | zeta-chain (TCR) associated protein kinase |
Gene Aliases: |
Srk, ZAP-70, mrtle, mur |
Location: |
Chr.1:36761798-36782821 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zap70 | NM_001289612.1 | 3 | 631 | NP_001276541.1 |
NM_001289765.1 | 10 | 1578 | NP_001276694.1 | |
NM_001289766.1 | 10 | 1543 | NP_001276695.1 | |
NM_009539.3 | 10 | 1535 | NP_033565.2 | |
AB083210.1 | 3 | 634 | BAC43746.1 | |
AB084383.1 | 3 | 473 | BAC67015.1 | |
AK039883.1 | 10 | 1472 | BAC30471.1 | |
AK089056.1 | 10 | 1507 | ||
BC029727.1 | 10 | 1493 | AAH29727.1 | |
U04379.1 | 10 | 1489 | AAA19250.1 | |
U77667.1 | 9 | 1381 | AAB36538.1 |