Assay Details
Target Gene Details
Entrez Gene ID: | 70827 |
Gene Name: | trafficking protein, kinesin binding 2 |
Gene Aliases: |
2900022D04Rik, 4733401O11Rik, AI480836, AW046271, Als2cr3, CALS-C, GRIF-1, GRIF1, OIP98, mKIAA0549 |
Location: |
Chr.1:58900449-58973482 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trak2 | NM_172406.3 | NP_765994.2 | ||
AK014647.1 | ||||
AK036650.1 | BAC29517.1 | |||
BC060681.1 | AAH60681.1 |
More Information
Set Membership: |
Intragenic Non-exonic |