Assay Details
Target Gene Details
Entrez Gene ID: | 215243 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
6030423D04Rik, AI115021, T3jam |
Location: |
Chr.1:193175504-193201546 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Traf3ip3 | NM_153137.4 | 2 | 311 | NP_694777.3 |
AK031400.1 | 2 | 251 | BAC27388.1 | |
AK036608.1 | 2 | 80 | ||
AK041994.1 | 2 | 312 | BAC31127.2 | |
AK080080.1 | 2 | 281 |
More Information
Set Membership: |
Intragenic Exonic |