Assay Details
Target Gene Details
Entrez Gene ID: | 56363 |
Gene Name: | transmembrane protein with EGF-like and two follistatin-like domains 2 |
Gene Aliases: |
4832418D20Rik, 7630402F16Rik, TR-2 |
Location: |
Chr.1:50909962-51187270 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmeff2 | NM_019790.4 | NP_062764.1 | ||
AB017270.1 | BAA87898.1 | |||
AB041565.1 | BAA95049.1 | |||
AK029307.1 | BAC26385.1 | |||
AK076254.1 | BAC36276.1 | |||
AK135038.1 | BAE22393.1 | |||
BC034850.1 | AAH34850.1 |
More Information
Set Membership: |
Intragenic Non-exonic |